Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11] (APC)

Supplier: STEMCELL Technologies
60063AZ 60063AZ.1
MSPP-60063AZEA 914.92 USD
MSPP-60063AZ MSPP-60063AZ1
Anti-SSEA-5 Mouse Monoclonal Antibody [clone: 8E11] (APC)
Antibodies
Mouse monoclonal IgG1 antibody against human SSEA-5, APC-conjugated.

  • Pluripotent stem cells

The 8e11 antibody reacts with stage-specific embryonic antigen-5 (SSEA-5), a terminal H type-1 glycan (Fuc1-2Galβ1-3GlcNAcβ1) expressed on the surface of cells in the inner cell mass of the human blastocyst during embryogenesis, and on undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells. Expression of SSEA-5 is rapidly down-regulated upon cellular differentiation. The 8e11 antibody can be used in conjunction with other pluripotency surface markers such as CD9 and CD90, or CD50 and CD200, to sort and remove undifferentiated teratoma-forming cells from incompletely differentiated stem cell cultures.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: SSEA-5
Clonality: Monoclonal
Clone: 8E11
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR