Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PE)

Supplier: STEMCELL Technologies
100-0431 60007PE 60007PE.1
MSPP-100-0431EA 2220.08 USD
MSPP-100-0431 MSPP-60007PE MSPP-60007PE1
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PE)
Antibodies
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD2, PE-conjugated.

  • T cells

The RPA-2.10 antibody reacts with CD2, an ~50 kDa type I transmembrane glycoprotein and a member of the immunoglobulin (Ig) superfamily; two Ig-like domains are located in its extracellular portion. CD2 is broadly expressed on peripheral T and NK cells, dendritic cells, erythrocytes, most thymic T cells, subsets of thymic B cells, and on the endothelium. CD2 expression appears early during T cell differentiation. Aberrant expression has been observed in some lymphomas and myeloid leukemias. CD2 is critically important for T cell activation and signaling, as well as lymphocyte adhesion. The primary ligand for CD2 is CD58 (LFA-3) located on antigen-presenting cells, with additional ligands comprising CD15 (SSEA-1), CD48 and CD59. Notably, the RPA-2.10 antibody blocks the mixed lymphocyte reaction.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Capuchin monkey, Chimpanzee, Cynomolgus monkey, Human, Pig, Pigtailed macaque, Rhesus monkey, Squirrel monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR