Anti-CD45.2 Mouse Monoclonal Antibody [clone: 104] (Biotin)
Supplier: STEMCELL Technologies
60118BT 60118BT.1
MSPP-60118BTEA 308.38 USD
MSPP-60118BT MSPP-60118BT1
Anti-CD45.2 Mouse Monoclonal Antibody [clone: 104] (Biotin)
Antibodies
Mouse monoclonal IgG2a antibody against mouse CD45.2, biotin-conjugated.
  • Granulocytes
  • Subsets
  • Hematopoietic Stem
  • Progenitor cells
  • Lymphocytes
  • Myeloid cells
This monoclonal antibody (clone 104) reacts with mouse alloantigen cluster of differentiation 45.2 (CD45.2), also known as Ly5.2 and leukocyte common antigen, a 180 -240 kD type I transmembrane glycoprotein. CD45.2 is an allelic form of CD45 expressed by all hematopoietic cells except mature erythrocytes and platelets in mouse strains 129, A, AKR, BALB/c, C3H/He, C57BL/6, C58, CBA, DBA/1, and DBA/2.More Product Information