Anti-CD4 Rat Monoclonal Antibody [clone: RM4-4] (APC)

Supplier: STEMCELL Technologies
60029AZ 60029AZ.1
MSPP-60029AZEA 517.38 USD
MSPP-60029AZ MSPP-60029AZ1
Anti-CD4 Rat Monoclonal Antibody [clone: RM4-4] (APC)
Antibodies
Rat monoclonal IgG2b antibody against mouse CD4, APC-conjugated.

  • T cells
  • T cells CD4
  • T cells Regulatory

The RM4-4 antibody reacts with mouse CD4, an ~55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobin (Ig) superfamily; CD4 contains four extracellular Ig-like domains. CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T-helper/inducer cells), and at lower levels on dendritic cells. In the mouse, CD4 is not expressed by monocytes/macrophages. CD4 binds to a non-polymorphic region of MHC II and acts as a co-receptor to the T cell receptor (TCR) in MHC II-restricted antigen recognition by enhancing the avidity of the association between the TCR and MHC II-antigen complex. CD4 also functions to amplify signals from the TCR to the cytoplasm through the interaction of its intracellular domain with cytoplasmic tyrosine kinases such as Lck. Binding of the RM4-4 antibody does not block binding of antibody clones RM4-5 or GK1.5.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-4
Conjugation: APC
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR