Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.7] (APC)
Supplier: STEMCELL Technologies
60023AZ 60023AZ.1
MSPP-60023AZEA 553.92 USD
MSPP-60023AZ MSPP-60023AZ1
Anti-CD8a Rat Monoclonal Antibody [clone: 53-6.7] (APC)
Antibodies
Rat monoclonal IgG2a antibody against mouse, toad CD8a, APC-conjugated.
  • T cells
  • T cells CD8
The 53-6.7 antibody reacts with murine CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8. CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα). CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell. It functions to strengthen the association between the TCR andMore Product Information