Anti-CD43 Mouse Monoclonal Antibody [clone: CD43-10G7] (PE)
Supplier: STEMCELL Technologies
60085PE 60085PE.1
MSPP-60085PEEA 536.38 USD
MSPP-60085PE MSPP-60085PE1
Anti-CD43 Mouse Monoclonal Antibody [clone: CD43-10G7] (PE)
Antibodies
Mouse monoclonal IgG1 antibody against human CD43, PE-conjugated.
  • B cells
The CD43-10G7 antibody reacts with human CD43, a type I transmembrane sialoglycoprotein expressed on the surface of most human hematopoietic cells, including thymocytes, peripheral T cells, NK cells, monocytes, granulocytes, some B cells (activated B cells and an early B cell subset in bone marrow), plasma cells, and hematopoietic progenitor cells. Low expression levels are observed on platelets. CD43 exhibits different glycoforms in the range of ~95 to 135 kDa, depending on theMore Product Information