Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631] (Alexa Fluor® 488)

Supplier: STEMCELL Technologies
60061AD 60061AD.1
MSPP-60061ADEA 914.92 USD
MSPP-60061AD MSPP-60061AD1
Anti-SSEA-3 Rat Monoclonal Antibody [clone: MC-631] (Alexa Fluor® 488)
Antibodies
Rat monoclonal IgM antibody against human, mouse, rat SSEA-3, Alexa Fluor® 488-conjugated.

  • Pluripotent stem cells

The MC-631 antibody reacts with a galactosylgloboside epitope on the stage-specific embryonic antigen-3 (SSEA-3), which is expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), undifferentiated embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey ES cell lines. No immunoreactivity is evident with undifferentiated mouse EC, EG and ES cells. Expression of SSEA-3 is down regulatedMore Product Information
Order Now

SPECIFICATIONS

Antigen Name SSEA-3
Environmentally Preferable
Clonality Monoclonal
Conjugation Alexa Fluor® 488
Clone MC-631
Reactivity Rhesus monkey,Human,Rat,Mouse
Host Rat
Gene ID 93961
Isotype IgM kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR