Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (FITC)

Supplier: STEMCELL Technologies
60004FI 60004FI.1
MSPP-60004FIEA 513 USD
MSPP-60004FI MSPP-60004FI1
Anti-CD14 Mouse Monoclonal Antibody [clone: M5E2] (FITC)
Antibodies
Mouse monoclonal IgG2a antibody against human, rhesus, cynomolgus CD14, FITC-conjugated.

  • Monocytes

The M5E2 antibody reacts with CD14, an ~53 - 55 kDa GPI-anchored transmembrane glycoprotein expressed at high levels on the surface of peripheral blood monocytes and macrophages, and at lower levels on granulocytes. An ~10-fold difference in expression levels between monocytes/macrophages and granulocytes makes CD14 a useful marker for distinguishing these cell populations. CD14 is also found on tissue macrophages, Langerhans cells and dendritic cells. CD14 functions as a high-affinity receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein and modulates LPS-dependent signal transduction during the immune response to gram-negative pathogens by acting as a co-receptor for TLR 4 and MD-2. This triggers activation of NF-kappa-B, cytokine secretion, and induction of the inflammatory response. Two soluble forms of CD14 have also been described (~48 and ~55 kDa). This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human Myeloid Positive Selection Kit (Catalog #18653) and EasySep™ Human CD14 Positive Selection Kit (Catalog #18058); partial blocking may be observed, as well as EasySep™ HLA Whole Blood CD33 Positive Selection Kit (Catalog #18287HLA), EasySep™ HLA Whole Blood Myeloid Positive Selection Kit (Catalog #18683HLA), and EasySep™ Human Buffy Coat CD14 Positive Selection Kit (Catalog #18088).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD14
Clonality: Monoclonal
Clone: M5E2
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Capuchin monkey, Chimpanzee, Common marmoset, Cotton-topped tamarin, Bovine, Cynomolgus monkey, Dog, Human, Macaque, Pig, Rhesus monkey, Sheep, Squirrel monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR