Anti-OCT4 (OCT3) Mouse Monoclonal Antibody [clone: 3A2A20]

Supplier: STEMCELL Technologies
60093 60093.1
MSPP-60093EA 793.62 USD
MSPP-60093 MSPP-600931
Anti-OCT4 (OCT3) Mouse Monoclonal Antibody [clone: 3A2A20]
Antibodies
Mouse monoclonal IgG2b antibody against human OCT4 (OCT3), unconjugated.

  • Pluripotent stem cells

This monoclonal antibody reacts with human octamer-binding transcription factor 4 (OCT4), also known as OCT3 and OCT3/4, an ~40 kDa homeodomain transcription factor belonging to the POU family, which is expressed in undifferentiated human embryonic stem (ES), induced pluripotent stem (iPS), embryonal carcinoma (EC), and embryonic germ (EG) cells. OCT4 binds to the octamer motif 5'-ATTTGCAT-3' and plays a key role in maintaining cells in a pluripotent stateMore Product Information
Order Now

SPECIFICATIONS

Antigen Name OCT4 (OCT3)
Environmentally Preferable
Clonality Monoclonal
Conjugation Unconjugated
Clone 3A2A20
Reactivity Human
Host Mouse
Gene ID 5460
Isotype IgG2b kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR