Anti-CD16 Mouse Monoclonal Antibody [clone: 3G8]

Supplier: STEMCELL Technologies
60041 60041.1
MSPP-600411EA 207.54 USD
MSPP-600411 MSPP-60041
Anti-CD16 Mouse Monoclonal Antibody [clone: 3G8]
Antibodies
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD16, unconjugated.

  • Granulocytes
  • Subsets
  • Monocytes

The 3G8 antibody reacts with CD16 (FcγRIII or low affinity IgG receptor III), a type 1 transmembrane glycoprotein belonging to the Ig superfamily. CD16 exists in two distinct isoforms: CD16a, a 50 - 65 kDa form expressed on NK cells, activated monocytes, macrophages, and placental trophoblasts, and CD16b, an ~48 kDa glycosylphosphatidylinositol (GPI)-anchored form expressed on neutrophils, basophils and eosinophils and found as at least two polymorphic variants, termed NA1 and NA2. CD16 binds weakly to the Fc region of monomeric, aggregated or complexed IgG, particularly the IgG1 and IgG3 isotypes. Binding of IgG to either CD16 isoform induces signaling pathways that modulate several types of responses, including antibody-dependent cell-mediated cytotoxicity (ADCC), phagocytosis, cytokine release and proliferation. CD16/IgG interactions can result in non-specific labeling in antibody-based detection and cell separation experiments and the 3G8 antibody may be employed as a blocking antibody to reduce non-specific binding.

Type:
Antigen: CD16
Clonality: Monoclonal
Clone: 3G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Baboon, Capuchin monkey, Chimpanzee, Common marmoset, Cotton-topped tamarin, Cynomolgus monkey, Human, Macaque, Rhesus monkey, Sooty mangabey, Squirrel monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR