Anti-Igg2b Isotype Control Mouse Monoclonal Antibody [clone: MPC-11] (PE)
Supplier: STEMCELL Technologies
60072PE 60072PE.1
MSPP-60072PEEA 285 USD
MSPP-60072PE MSPP-60072PE1
Anti-Igg2b Isotype Control Mouse Monoclonal Antibody [clone: MPC-11] (PE)
Antibodies
Mouse monoclonal IgG2b, kappa isotype control antibody, PE-conjugated.
The MPC-11 antibody (IgG2b, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of an appropriateMore Product Information