Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (APC)

Supplier: STEMCELL Technologies
100-0467 60018AZ.1
MSPP-100-0467EA 1460.08 USD
MSPP-100-0467 MSPP-60018AZ1
Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (APC)
Antibodies
Mouse monoclonal IgG1 antibody against human, chimpanzee CD45, APC-conjugated.

  • Granulocytes
  • Subsets
  • Hematopoietic stem
  • Progenitor cells
  • Leukocytes
  • Lymphocytes
  • Myeloid cells

The HI30 antibody reacts with all isoforms of CD45, a type I transmembrane glycoprotein expressed on the surface of most hematopoietic cells except mature erythrocytes, platelets, and plasma cells; expression of CD45 is lost during differentiation of these cell types. CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation. The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains (one of which is enzymatically active) that are involved in modulating the function of intracellular substrates such as the Src kinases Lck and Fyn. Several isoforms of CD45 have been identified, the expression of which differs according to cell type and functional status. Alternative splicing of three exons (4, 5, 6) encoding the extracellular RA, RB, and RC polypeptide sequences gives rise to up to 8 isoforms with molecular masses in the range of 180 - 240 kDa. The leukocyte common antigen (LCA), the region recognized by the HI30 antibody, is an extracellular region located proximal to the membrane and common to all isoforms of CD45. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human T Cell Enrichment Kit (Catalog #19051), EasySep™ Human CD4+ T Cell Enrichment Kit (Catalog #19052) and EasySep™ Human CD45 Depletion Kit (Catalog #18259; partial blocking may be observed).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD45
Clonality: Monoclonal
Clone: HI30
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR