Mouse Recombinant TREM2 (His & AVI Tag), (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
50149-M49HB100 50149-M49H-B20
77444-674EA 688 USD
77444-674 77444-676
Mouse Recombinant TREM2 (His & AVI Tag), (from HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the Mouse TREM2 (NP_112544.1)(Met1-Ser171) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.