Recombinant eRFP (His tag) (from E. coli)
Supplier: Sino Biological
R620-30BH-50 R620-30BH-20
77563-408EA 336 USD
77563-408 77563-406
Recombinant eRFP (His tag) (from E. coli)
Proteins and Peptides
Recombinant full length Bubble-tip anemone eRFP was expressed in E. coli cells using an N-terminal His tag.
  • Shipping temperature: Dry ice