Mouse Recombinant RIPK1 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
R07M-11G-10 R07M-11G-50 R07M-11G-500
77562-978EA 378 USD
77562-978 77562-980 77562-982
Mouse Recombinant RIPK1 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant mouse RIPK1 (1-327) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice