Human Recombinant HRAS (G12V, His tag) (from E. coli)
Supplier: Sino Biological
R05-32H-100 R05-32H-1000 R05-32H-20
77562-824EA 448 USD
77562-824 77562-826 77562-828
Human Recombinant HRAS (G12V, His tag) (from E. coli)
Proteins and Peptides
Recombinant human RAS1 (2-186, G12V) was expressed in E.coli cells using an N-terminal His tag. The HRAS1(G12V) mutant is constitutively active.
  • Shipping temperature: Dry ice