Human Recombinant HRAS (G12A, His-TEV tag) (from E. coli)
Supplier: Sino Biological
R05-32ATH-100 R05-32ATH-1000 R05-32ATH-20
77562-812EA 578 USD
77562-812 77562-814 77562-816
Human Recombinant HRAS (G12A, His-TEV tag) (from E. coli)
Proteins and Peptides
Recombinant human HRAS (G12A) (2-186) was expressed in E. coli cells using an N-terminal His-TEV tag. HRAS (G12A) is constitutively active.
  • Shipping temperature: Dry ice