Human Recombinant PTPRB (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P49-21G-10 P49-21G-50 P49-21G-500
77561-924EA 268 USD
77561-924 77561-926 77561-928
Human Recombinant PTPRB (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human PTPRB (1552-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice