Human Recombinant PLK3 (His tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P43-13H-10 P43-13H-50 P43-13H-500
77561-830EA 348 USD
77561-830 77561-832 77561-834
Human Recombinant PLK3 (His tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human PLK3 (57-340) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
  • Shipping temperature: Dry ice