Human Recombinant HDAC11 (GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
H93-30G-10 H93-30G-50 H93-30G-500
77558-382EA 378 USD
77558-382 77558-384 77558-386
Human Recombinant HDAC11 (GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length human HDAC11 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice