Human Recombinant H11 (HSP22, GST tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
H09-34G-100 H09-34G-1000 H09-34G-20
77558-070EA 818 USD
77558-070 77558-072 77558-074
Human Recombinant H11 (HSP22, GST tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length human H11 (HSP22) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice