Human Recombinant GRB2 (SH3-1 domain, GST tag) (from E. coli)
Supplier: Sino Biological
G45-351G-100 G45-351G-1000 G45-351G-20
77557-982EA 818 USD
77557-982 77557-984 77557-986
Human Recombinant GRB2 (SH3-1 domain, GST tag) (from E. coli)
Proteins and Peptides
Recombinant human GRB2 SH3-1 domain (2-64) was expressed in E. coli cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice