Human Recombinant FGFR3 (d613-653, GST tag), Unactive (from Baculovirus (Insect cells))
Supplier: Sino Biological
F06-16HG-100 F06-16HG-1000 F06-16HG-20
77557-560EA 818 USD
77557-560 77557-562 77557-564
Human Recombinant FGFR3 (d613-653, GST tag), Unactive (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human FGFR3 (d613-653) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice