Human Recombinant 4EBP1 (GST tag) (from E. coli)
Supplier: Sino Biological
E35-54G-100 E35-54G-1000 E35-54G-20
77557-298EA 818 USD
77557-298 77557-300 77557-302
Human Recombinant 4EBP1 (GST tag) (from E. coli)
Proteins and Peptides
Recombinant full length human 4EBP1 was expressed in E. coli cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice