Human Recombinant HER2 (G776VC, GST tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
E27-132EG-10 E27-132EG-50 E27-132EG-500
77557-178EA 338 USD
77557-178 77557-180 77557-182
Human Recombinant HER2 (G776VC, GST tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant human HER2 (G776VC) (676-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice