Human Recombinant TRKA (F589L, GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
N16-12EG-10 N16-12EG-50 N16-12EG-500
77560-618EA 338 USD
77560-618 77560-620 77560-622
Human Recombinant TRKA (F589L, GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human TRKA (F589L) (440 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice