Human Recombinant MET (R970C, GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M52-12OG-10 M52-12OG-50 M52-12OG-500
77560-230EA 338 USD
77560-230 77560-232 77560-234
Human Recombinant MET (R970C, GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human MET (R970C) (956 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice