Human Recombinant p38 Alpha (K53A, His Tag), Unactive (from E. coli)
Supplier: Sino Biological
M39-16H-100 M39-16H-1000 M39-16H-20
77559-918EA 818 USD
77559-918 77559-920 77559-922
Human Recombinant p38 Alpha (K53A, His Tag), Unactive (from E. coli)
Proteins and Peptides
Recombinant full length human mutant p38 alpha (K53A) was expressed in E. coli using an N to terminal His tag. This kinase to dead protein is designed for up to stream kinase assay.
  • Shipping temperature: Dry ice