Human Recombinant MEK3 (MAP2K3, GST Tag), Active (from Baculovirus (Insect cells))
Supplier: Sino Biological
M04-10G-10 M04-10G-50 M04-10G-500
77559-414EA 348 USD
77559-414 77559-416 77559-418
Human Recombinant MEK3 (MAP2K3, GST Tag), Active (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length Human MEK3 was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice