Human Recombinant LATS2 (GST Tag), Unactive (from Baculovirus (Insect cells))
Supplier: Sino Biological
L02-35G-100 L02-35G-1000 L02-35G-20
77559-132EA 818 USD
77559-132 77559-134 77559-136
Human Recombinant LATS2 (GST Tag), Unactive (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human LATS2 (480 to 1088) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice