Human Recombinant JAK1 (d966-989, GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
J01-12DG-10 J01-12DG-50 J01-12DG-500
77558-626EA 338 USD
77558-626 77558-628 77558-630
Human Recombinant JAK1 (d966-989, GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human JAK1 (d966 to 989) (438 to end) was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.
  • Shipping temperature: Dry ice