Human Recombinant Cyclin T2 (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C86-30BG-20 C86-30BG-100 C86-30BG-1000
77555-888EA 238 USD
77555-888 77555-884 77555-886
Human Recombinant Cyclin T2 (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human Cyclin T2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice