Human Recombinant CDK9/CyclinK (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C40-10G-50 C40-10G-500
77555-540EA 1168 USD
77555-540 77555-542
Human Recombinant CDK9/CyclinK (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human CDK9 and CyclinK were co-expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
  • Shipping temperature: Dry ice