Human Recombinant CDK13/CyclinK (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C25-11BG-10 C25-11BG-50 C25-11BG-500
77555-254EA 378 USD
77555-254 77555-256 77555-258
Human Recombinant CDK13/CyclinK (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human CDK13 (516-end) /Cyclin K complex was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag on both proteins.
  • Shipping temperature: Dry ice