Human Recombinant CDC25A (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
C04-20G-10 C04-20G-50 C04-20G-500
77554-956EA 328 USD
77554-956 77554-958 77554-960
Human Recombinant CDC25A (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human CDC25A was expressed in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice