Human Recombinant BRAF (D594G, GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
B08-12MG-10 B08-12MG-50 B08-12MG-500
77554-710EA 338 USD
77554-710 77554-712 77554-714
Human Recombinant BRAF (D594G, GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant human BRAF (D594G) (381-end) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice