Human Recombinant AKT3 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A18-10G-500 A18-10G-10 A18-10G-50
77554-148EA 2638 USD
77554-148 77554-144 77554-146
Human Recombinant AKT3 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human AKT3 was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag.
  • Shipping temperature: Dry ice