Mouse Recombinant AATK (GST tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
A02-35G-100 A02-35G-1000 A02-35G-20
77553-744EA 818 USD
77553-744 77553-746 77553-748
Mouse Recombinant AATK (GST tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant mouse AATK (8-411) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
  • Shipping temperature: Dry ice