Antipain dihydrochloride
Supplier: RPI
A20560-0.005
A20560-0.025
76344-812EA
65.99
USD
76344-812
77510-170
Antipain dihydrochloride
Proteins and Peptides
Reversible cysteine and serine protease inhibitor of cathepsins A and B, papain and trypsin.
- Appearance: White powder
- Solubility (0.01%, Water): Pass
- 1-H NMR: Conform to structure
- Purity (HPLC): ≥97.0%
- Element analysis (C): 47.86% (theoretical)
- Element analysis (H): 6.84% (theoretical)
- Element analysis (N): 20.67% (theoretical)
- Element analysis (Cl): 10.46% (theoretical)
- Specific optical rotation [a]20D (c=1.0, H₂O): Report °
- Biochemical activity: Report KU/mg
- BSE/TSE free: BSE/TSE free
- Country of origin: Report
- Working concentration: 1 to 100uM
Caution: Research or further manufacturing use only, not for food or drug use.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...