Colistin sulfate
Supplier: RPI
C70700-1.0 C70700-5.0 C70700-10.0
76345-644EA 126.92 USD
76345-644 76345-646 77510-228
Colistin sulfate
Proteins and Peptides
Colistin sulfate is a potent antibiotic that has gained significant importance in the life science marketplace. It belongs to the polymyxin class of antibiotics and is primarily used to combat infections caused by multidrug-resistant Gram-negative bacteria, including strains resistant to other antibiotics.
  • BSE/TSE Free
  • Country of Origin: China
Colistin sulfate is effective against bacteria such as Pseudomonas aeruginosa, Acinetobacter baumannii, and Klebsiella pneumoniae, which are known to be highly resistant to multiple antibiotics.
Additionally, It is widely employed in microbiology laboratories for testing bacterial susceptibility to antibiotics, determining minimum inhibitory concentrations (MIC), and conducting antimicrobial susceptibility tests.
GHS Symbol: GHS06
Hazard Class: 6.1
Packing Group: III
Signalword: Danger