Calbiochem® Bovine Hemoglobin (from Erythrocytes)
Supplier: MilliporeSigma
3745-5GM 374834-10GM
80052-178EA 153.64 USD
80052-178 EM374834-10GM
Calbiochem® Bovine Hemoglobin (from Erythrocytes)
Proteins and Peptides
A major oxygen-transporting component of red blood cells. A nitric oxide scavenger. Blocks carbachol-stimulated cGMP production. This preparation contains primarily ferric-hemoglobin and must be reduced to the ferrous form to bind molecular oxygen.