Human Recombinant RNASET2 Protein (His Tag) (From Baculovirus-Insect Cells)
Supplier: Sino Biological
13509-H08B-50
77447-442EA 298 USD
77447-442
Human Recombinant RNASET2 Protein (His Tag) (From Baculovirus-Insect Cells)
Proteins and Peptides
A DNA sequence encoding the human RNASET2 isoform 1 (O00584-1) (Met 1-His 256) was fused with a polyhistidine tag at the C-terminus.