Human Recombinant EphA7/EHK3 Protein (His Tag) (From HEK293 Cells)
Supplier: Sino Biological
11657-H08H-1
77447-150EA 1938 USD
77447-150
Human Recombinant EphA7/EHK3 Protein (His Tag) (From HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human EPHA7 (NP_004431.1) extracellular domain (Met 1-Ile 556) was fused with a polyhistidine tag at the C-terminus.