Human Recombinant CD30/TNFRSF8 Protein (His Tag) (From HEK293 Cells), Biotinylated
Supplier: Sino Biological
10777-H08H-B-100 10777-H08H-B-20
77446-798EA 198 USD
77446-798 77446-796
Human Recombinant CD30/TNFRSF8 Protein (His Tag) (From HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the Human TNFRSF8 (NP_001234.2) (Met1-Lys379) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.