Human Recombinant B7-DC/PD-L2/CD273 Protein (His & AVI Tag) (From HEK293 Cells), Biotinylated
Supplier: Sino Biological
10292-H27H-B-100 10292-H27H-B-20
77446-468EA 298 USD
77446-468 77446-466
Human Recombinant B7-DC/PD-L2/CD273 Protein (His & AVI Tag) (From HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the human PDCD1LG2 (NP_079515.2) (Met1-Pro219) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.