Human Recombinant CD84 Protein (His Tag) (From HEK293 Cells)
Supplier: Sino Biological
10100-H08H-1
77446-292EA 1418 USD
77446-292
Human Recombinant CD84 Protein (His Tag) (From HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Gly 225) of human CD84 (NP_003865.1) was expressed with a C-terminal polyhistidine tag.