Human Recombinant 4-1BB/CD137 Protein (ECD, AVI & His Tag) (From HEK293 Cells), Biotinylated
Supplier: Sino Biological
10041-H27H-B-100 10041-H27H-B-20
77446-254EA 688 USD
77446-254 77446-256
Human Recombinant 4-1BB/CD137 Protein (ECD, AVI & His Tag) (From HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the human TNFRSF9 (NP_001552.2) (Met1-Gln186) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.