Human Recombinant CRP/C-Reactive Protein (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
11250-HNAH-B-20 11250-HNAH-B-100
77447-060EA 198 USD
77447-060 77447-058
Human Recombinant CRP/C-Reactive Protein (from HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the human CRP (P02741-1) (Met1-Pro224) was expressed and purified. The purified protein was biotinylated in vitro.