Human Recombinant TIM-3 Protein (Fc & AVI Tag) (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
10390-H41H-B-100 10390-H41H-B-20
77446-548EA 688 USD
77446-548 77446-550
Human Recombinant TIM-3 Protein (Fc & AVI Tag) (from HEK293 Cells), Biotinylated
Proteins and Peptides
A DNA sequence encoding the human HAVCR2 (NP_116171.3)(Met1-Arg200) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.