Human Recombinant TLR3 Protein (His Tag) (from Baculovirus-Insect Cells)
Supplier: Sino Biological
10190-H08B-1 10190-H08B-100
77446-388EA 2268 USD
77446-388 77446-390
Human Recombinant TLR3 Protein (His Tag) (from Baculovirus-Insect Cells)
Proteins and Peptides
A DNA sequence encoding the Human TLR3 (AAH59372.1) (Lys27-Ser711) was expressed with a polyhistidine tag at the C-terminus.